Tetraethylammonium bromide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.22147.0250 8.22147.1000 8.22147.0250 8.22147.1000
EM8.22147.0250CS 706.26 CAD
EM8.22147.0250 EM8.22147.1000 CA8.22147.0250 CA8.22147.1000
Tetraethylammonium bromide for synthesis, Sigma-Aldrich®
Tetraethylammonium bromide
Formula: C₈H₂₀BrN
Melting Pt: > 285 °C
Density: 1.43 g/cm³ (20 °C)
MDL Number: MFCD00011825
CAS Number: 71-91-0
EINECS: 200-769-4
Merck Index: 13,09274

Order Now

Specification Test Results

Solubility 2795 g/l (25 °C)
Melting Point 285 °C
Molar Mass 210.16 g/mol
Density 1.397 g/cm3 (20 °C)
pH 6.5 (100 g/l, H2O, 20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR