1,2,4-Trichlorobenzene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.21152.1000 8.21152.2500 8.21152.1000 8.21152.2500
EM8.21152.0100CS 1118.02 CAD
EM8.21152.0100 EM8.21152.1000 EM8.21152.2500 CA8.21152.0100 CA8.21152.1000 CA8.21152.2500
1,2,4-Trichlorobenzene for synthesis, Sigma-Aldrich®
1,2,4-Trichlorobenzene
Formula: C₆H₃Cl₃
Boiling Pt: 214.4 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.45 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: 6.1,III
Merck Index: 13,09704

Order Now

Specification Test Results

Solubility 0.049 g/l (20 °C)
Melting Point 17 °C
Molar Mass 181.45 g/mol
Boiling Point 213.5 °C (1013 hPa)
Vapor Pressure 1.3 hPa (20 °C)
Flash Point 110 °C
Explosion Limit 2.5 - 6.6 %(V)
Density 1.45 g/cm3 (20 °C)
Ignition Point 571 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR