Triethyl orthoacetate, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20934.0050
CA8.20934.0050EA 40 CAD
CA8.20934.0050
Triethyl orthoacetate, Sigma-Aldrich®
Triethyl orthoacetate
Formula: C₈H₁₈O₃
Boiling Pt: 142 °C (1013 hPa)
Density: 0.8847 g/cm³ (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00009223
CAS Number: 78-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR