Di-tert-butyl peroxide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20248.0250 8.20248.0250
EM8.20248.0250CS 2870.86 CAD
EM8.20248.0250 CA8.20248.0250
Di-tert-butyl peroxide for synthesis, Sigma-Aldrich®
Di-tert-butyl peroxide
Formula: C₈H₁₈O₂
MDL Number: MFCD00008803
CAS Number: 110-05-4
UN: 3107
ADR: 5.2,

Order Now

Specification Test Results

Solubility 0.063 g/l (20 °C)
Molar Mass 146.23 g/mol
Boiling Point 110 - 115 °C
Vapor Pressure 1000 hPa (110 °C)
Flash Point 6 °C
Density 0.80 g/cm3 (20 °C)
Ignition Point 182 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR