Citric acid, anhydrous for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18707.1000 8.18707.5000 8.18707.1000 8.18707.5000
EM8.18707.1000CS 922.72 CAD
EM8.18707.1000 EM8.18707.5000 CA8.18707.1000 CA8.18707.5000
Citric acid, anhydrous for synthesis, Sigma-Aldrich®
Citric acid
Formula: HOC(COOH)(CH₂COOH)₂
MDL Number: MFCD00011669
CAS Number: 77-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (alkalimetric) ≥ 99 %
Identity (IR-spectrum) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR