Chloramine-T (sodium salt) trihydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18705.0005 8.18705.0250 8.18705.1000 8.18705.0005 8.18705.0250 8.18705.1000
EM8.18705.0005CS 66032.94 CAD
EM8.18705.0005 EM8.18705.0250 EM8.18705.1000 CA8.18705.0005 CA8.18705.0250 CA8.18705.1000
Chloramine-T (sodium salt) trihydrate for synthesis, Sigma-Aldrich®
Chloramine T (sodium salt) trihydrate
Formula: CH₃C₆H₄SO₂NClNa·3H₂O
MDL Number: MFCD00149066
CAS Number: 7080-50-4
UN: 2928
ADR: 6.1 (8),II

Order Now

Specification Test Results

Solubility 150 g/l (25 °C)
Molar Mass 281.69 g/mol
Bulk Density 540 - 680 kg/m3
Flash Point 192 °C
pH 8 - 10 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR