(S)-(-)-β-Citronellol, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.18378.0010
CA8.18378.0010EA 127 CAD
CA8.18378.0010
(S)-(-)-β-Citronellol, Sigma-Aldrich®
(S)-(-)-β-Citronellol
Formula: C₁₀H₂₀O
Density: 0.855 g/cm³ (20 °C)
MDL Number: MFCD00063214
CAS Number: 7540-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR