Polyvinyl alcohol, M.W. ∼ 145000 zS, hydrolysed_fully, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

MQ200

8.14894.0101 8.14894.1001 8.14894.0101 8.14894.1001
EM8.14894.0101CS 886.38 CAD
EM8.14894.0101 EM8.14894.1001 CA8.14894.0101 CA8.14894.1001
Polyvinyl alcohol, M.W. ∼ 145000 zS, hydrolysed_fully, Sigma-Aldrich®
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Melting Point 160 - 240 °C
Bulk Density 400 - 600 kg/m3
Density 0.4 - 0.6 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR