Bis (8-hydroxyquinolinium) sulfate monohydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14528.0100 8.14528.0100
EM8.14528.0100CS 673.08 CAD
EM8.14528.0100 CA8.14528.0100
Bis (8-hydroxyquinolinium) sulfate monohydrate for synthesis, Sigma-Aldrich®
Bis (8-hydroxyquinolinium) sulphate monohydrate
Formula: C₁₈H₁₆N₂O₆S·1H₂O
MDL Number: MFCD00149610
CAS Number: 207386-91-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 177 °C
Molar Mass 406.41 g/mol
pH 3.2 (H2O, 20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR