Bis(4-hydroxyphenyl) sulfide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 4,4'-Thiodiphenol

M-Clarity™ quality level = MQ200

8.14209.0100 8.14209.0100
EM8.14209.0100CS 3156.84 CAD
EM8.14209.0100 CA8.14209.0100
Bis(4-hydroxyphenyl) sulfide for synthesis, Sigma-Aldrich®
Bis(4-hydroxyphenyl) sulphide
Formula: C₁₂H₁₀O₂S
MW: 218.28 g/mol
MDL Number: MFCD00002349
CAS Number: 2664-63-3
UN: 3261

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 149 - 152 °C
Molar Mass 218.26 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR