trans-Stilbene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.14152.0100
CA8.14152.0100EA 219 CAD
CA8.14152.0100
trans-Stilbene for synthesis, Sigma-Aldrich®
trans-Stilbene
Formula: C₁₄H₁₂
MDL Number: MFCD00064300
CAS Number: 103-30-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52acf17325a810eecda8f5a4d795a5b31a1f6434&vid=6d95185bf3c5a4ec46f2df78329eb0b669b2b255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.00029 g/l (25 °C)
Melting Point 121 - 124 °C
Molar Mass 180.24 g/mol
Boiling Point 306 - 307 °C (1013 hPa)
Density 1.14 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR