Zinc(II) acetylacetonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')zinc (II), Zinc acetylacetonate, Zinc (II) 2,4-pentanedionate, Zinc (II) bis(2,4-pentanedionato-O,O')

M-Clarity™ quality level = MQ200

8.08803.0100
CA8.08803.0100CS 1810 CAD
CA8.08803.0100
Zinc(II) acetylacetonate for synthesis, Sigma-Aldrich®
Zinc (II) acetylacetonate
Formula: C₁₀H₁₄O₄Zn
MW: 263.61 g/mol
MDL Number: MFCD00000035
CAS Number: 14024-63-6

Order Now

Specification Test Results

Solubility 6.9 g/l (20 °C)
Melting Point 124 - 126 °C
Molar Mass 263.61 g/mol
Bulk Density 500 kg/m3
Boiling Point 129 - 131 °C (13 hPa)
Vapor Pressure 1 hPa (20 °C)
pH 10 (4 g/l, H2O, 20 °C)
Ignition Point 580 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR