DL-Tropic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 3-Hydroxy-2-phenylpropanoic acid, Tropic acid, 3-Hydroxy-2-phenylpropionic acid, (±)-Tropic acid, 2-Phenylhydracrylic acid

M-Clarity™ quality level = MQ200

8.08384.0025
CA8.08384.0025EA 120 CAD
CA8.08384.0025
DL-Tropic acid for synthesis, Sigma-Aldrich®
DL-Tropic acid
Formula: C₉H₁₀O₃
MDL Number: MFCD00004255
CAS Number: 529-64-6

Order Now

Specification Test Results

Solubility 20 g/l (20 °C)
Melting Point 115 - 118 °C
Molar Mass 166.17 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR