2-Propanethiol 98,0 GC_ASSAY_METHOD zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Isopropyl mercaptan, Isopropanethiol, 2-Mercaptopropane

MQ200

8.07532.0100 8.07532.0500 8.07532.9045
CA8.07532.0100CS 2331 CAD
CA8.07532.0100 CA8.07532.0500 CA8.07532.9045
2-Propanethiol 98,0 GC_ASSAY_METHOD zS, Sigma-Aldrich®
2-Propanethiol
Formula: C₃H₈S
MW: 76.16 g/mol
Boiling Pt: 57
Melting Pt: -130
Density: 0.813 g/cm3 at 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00004863
CAS Number: 75-33-2
UN: 2402
ADR: 3 II

Order Now

Specification Test Results

Solubility (20°C) slightly Soluble
Melting Point -130 °C
Boiling Point 51 - 53 °C (1013 hPa)
Vapor Pressure 605 hPa (37 °C)
Flash Point -34 °C
Refractive Index 1.4255 (20 °C, 589 nm)
Density 0.81 g/cm3 (20 °C)
Synonyms Isopropyl mercaptan
Hill Formula C3H8S
Chemical Formula (CH3)2CHSH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR