Dibutyltin oxide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.03539.0005 8.03539.0250
CA8.03539.0005CS 115157 CAD
CA8.03539.0005 CA8.03539.0250
Dibutyltin oxide for synthesis, Sigma-Aldrich®
Dibutyltin oxide
Formula: C₈H₁₈OSn
Melting Pt: > 300 °C
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001992
CAS Number: 818-08-6
EINECS: 212-449-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility 0.004 g/l (20 °C)
Molar Mass 248.94 g/mol
Bulk Density 650 - 750 kg/m3
Boiling Point 300 °C decomposes
Density 1.5 g/cm3 (20 °C)
Ignition Point 525 °C
Hazard Class 9
Hazard Class Description Miscellaneous hazardous material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR