N,N-Dimethyl-1,3-diaminopropane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.03503.0100 8.03503.1000 8.03503.9035
CA8.03503.0100CS 597 CAD
CA8.03503.0100 CA8.03503.1000 CA8.03503.9035
N,N-Dimethyl-1,3-diaminopropane for synthesis, Sigma-Aldrich®
N,N-Dimethyl-1,3-diaminopropane
Formula: C₅H₁₄N₂
Boiling Pt: 132…140 °C (1013 hPa)
Melting Pt: <–50 °C
Density: 0.82 g/cm3 at 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008216
CAS Number: 109-55-7
UN: 2734
ADR: 8.3,II

Order Now

Specification Test Results

Solubility (20°C) Soluble
Boiling Point 132 - 140 °C (1013 hPa)
Vapor Pressure 17 hPa (30 °C)
Flash Point 32 °C
Refractive Index 1.4415 (20 °C, 589 nm)
Explosion Limit 2.3 - 12.35 %(V)
Density 0.82 g/cm3 (20 °C)
pH 12.7 (100 g/l, H2O, 20 °C)
Ignition Point 215°C
Synonyms 3-Dimethylaminopropylamine, N,N-Dimethyl-1,3-propanediamine, N,N-Dimethyl-1,3-diaminopropane
Hill Formula C5H14N2
Chemical Formula (CH3)2N(CH2)3NH2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR