Benzamide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Benzoic acid amide

M-Clarity™ quality level = MQ200

8.02191.9025
CA8.02191.9025EA 1653 CAD
CA8.02191.9025
Benzamide for synthesis, Sigma-Aldrich®
Benzamide
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 288
Melting Pt: 124
Density: 1.34 g/cm3 (20 °C)
Flash Pt: 180
Storage Temperature: Ambient
MDL Number: MFCD00007968
CAS Number: 55-21-0
Merck Index: 13,01059

Order Now

Specification Test Results

Solubility 13 g/l (25°C)
Melting Point 124 - 127 °C
Molar Mass 550 kg/m3
Bulk Density 550 kg/m3
Vapor Pressure 0.000056 hPa (20 °C)
Flash Point 180 °C
Density 1.34 g/cm3 (20 °C)
pH 6.9 (H2O) (saturated solution)
Ignition Point 500°C
Synonyms Benzoic acid amide
Hill Formula C7H7NO
Chemical Formula C6H5CONH2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR