Ethylenediamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00947.0100 8.00947.0500 8.00947.1000 8.00947.2500 8.00947.9051
CA8.00947.0100CS 1390 CAD
CA8.00947.0100 CA8.00947.0500 CA8.00947.1000 CA8.00947.2500 CA8.00947.9051
Ethylenediamine for synthesis, Sigma-Aldrich®
Ethylenediamine
Formula: C₂H₈N₂
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: 8.5 °C
Density: 0.898 g/cm³ (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8 (3),II
Merck Index: 13,03829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cbd90391008b8df01337e52ed5760383c063baa&vid=9a49702fe7150a0d3cd56afb586972447ecb6f85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 11 °C
Molar Mass 60.10 g/mol
Boiling Point 116 °C (1013 hPa)
Vapor Pressure 12 hPa (20 °C)
Flash Point 38 °C
Explosion Limit 2.5 - 16.3 %(V)
Density 0.897 g/cm3 (20 °C)
pH 12.2 (100 g/l, H2O, 20 °C)
Ignition Point 405 °C
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR