N-Ethyldiisopropylamine zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA, Ethyldiisopropylamine

MQ200

8.00894.1000
CA8.00894.0250CS 3026.7 CAD
CA8.00894.0250 CA8.00894.1000 CA8.00894.2500
N-Ethyldiisopropylamine zS, Sigma-Aldrich®
N-Ethyldiisopropylamine
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 127
Melting Pt: -50
Density: 0.76 g/cm³ (20 °C)
Flash Pt: 6
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
EINECS: 230-392-0
UN: 2733
ADR: 3 II

Order Now

Specification Test Results

Solubility 3.9 g/l (20 °C)
Melting Point -46 °C
Molar Mass 129.24 g/mol
Boiling Point 127 °C (1013 hPa)
Vapor Pressure 16 hPa (20 °C)
Flash Point 9.5 °C
Refractive Index 1.4121 (25 °C, 589 nm)
Explosion Limit 0.7 - 6.3 %(V)
Density 0.76 g/cm3 (20 °C)
pH 12.3 (H2O) (as an emulsion)
Ignition Point 240 °C
Viscosity kinematic 0.88 mm2/s (20 °C)
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR