Tetrachlorophthalic anhydride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00741.0100
CA8.00741.0100CS 2184 CAD
CA8.00741.0100
Tetrachlorophthalic anhydride for synthesis, Sigma-Aldrich®
Tetrachlorophthalic anhydride
Formula: C₈Cl₄O₃
MW: 285.9 g/mol
Boiling Pt: 371
Density: 1.52 g/cm³ (275 °C)
MDL Number: MFCD00005920
CAS Number: 117-08-8
EINECS: 204-171-4
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 253 - 256 °C
Molar Mass 285.89 g/mol
Boiling Point 350 °C (1.013 hPa)
Vapor Pressure 0.0021 kPa (145 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR