p-Anisidine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00458.0100 8.00458.0500
CA8.00458.0100EA 60 CAD
CA8.00458.0100 CA8.00458.0500
p-Anisidine for synthesis, Sigma-Aldrich®
p-Anisidine
Formula: C₇H₉NO
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 55…58 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00007864
CAS Number: 104-94-9
EINECS: 203-254-2
UN: 3143
ADR: 6.1,III
Merck Index: 13,00670

Order Now

Specification Test Results

Solubility 21 g/l (20 °C)
Melting Point 55 - 58 °C
Molar Mass 123.15 g/mol
Bulk Density 700 kg/m3
Boiling Point 243 °C
Vapor Pressure 0.02 hPa (20 °C)
Flash Point 122 °C
Density 1.06 g/cm3 (63 °C)
pH 7.7 (1 g/l, H2O, 20 °C)
Ignition Point 450 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR