ChiraDex HPLC Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
79925CB-584
CA79925CB-584EA 2856.01 CAD
CA79925CB-584
ChiraDex HPLC Columns, Agilent Technologies
Chromatography Columns
ChiraDex columns allow for direct racemic separations without derivatization.
  • Cartridge columns
  • Novel manufacturing process bonds ß-cyclodextrin to spherical 5 µm silica gel by means of a chemical spacer
Applications include barbiturates, benzothiadiazines, calcium antagonists, and PTH-amino acids.
Note: Cartridge columns require reusable cartridge hardware (101481-418).