Slide Drying/Draining Holder, JAC Medical Products
Supplier: JAC
JAC22
CA78010-056EA 93.63 CAD
CA78010-056
Slide Drying/Draining Holder, JAC Medical Products
Slide Holders
Ideal for bone marrow smears, this holder accommodates 12 slides individually or 24 slides back-to-back
Holder provides protection from breakage and contamination of slides. Openings in the well bottoms facilitate draining. Constructed of thick, high-quality polypropylene for durability. Autoclavable.