Water Distiller, 9000 Model, Heidolph
Supplier: Heidolph NA, LLC
023211145
CA77889-068EA 1340.25 CAD
CA77889-068
Water Distiller, 9000 Model, Heidolph
Distillation Systems Automatic Distillation Systems
Produces 1 gallon of 100% steam distilled water in 4 hours (up to 6 gallons per day), reduces 99% of total dissolved solids (TDS), and has a dripless stop-and-serve feature.