Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
AP-4500 AP-4408 AP-4308 AP-4309 AP-4310 AP-4225 AP-4521 AP-4219 AP-4302 AP-4303 AP-4501 AP-4502 AP-4549 AP-4548 AP-4436 AP-4438 AP-4517 AP-4518 AP-4528 AP-4522 AP-4793 AP-4794 AP-4787 AP-4788 AP-4790 AP-4791 AP-4792 AP-4796 AP-4797 AP-4910 AP-4912 AP-4913 AP-4914 AP-4915 AP-4916 AP-4917 AP-4918 AP-4919 AP-4920 AP-4921 AP-4438TC
CA16003-720PK 3836 CAD
CA16003-720 CA16003-722 CA16003-724 CA16003-726 CA16003-728 CA16003-730 CA28143-384 CA28143-392 CA28143-866 CA28143-869 CA28143-870 CA28143-872 CA28143-884 CA28143-886 CA28143-910 CA28143-914 CA28143-274 CA28143-882 CA28143-901 CA28143-903 CA28143-912 28143-278 CA97027-950 CA97027-956 CA97027-938 CA97027-940 CA97027-944 CA97027-946 CA97027-948 CA97027-962 CA97027-964 76308-662 76308-664 76308-666 76308-668 76308-670 76308-672 76308-674 76308-676 76308-678 76308-680 76308-682 76308-684 CA77610-708
Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Filters Syringe Filters
The new Acrodisc® One and Acrodisc® PSF premium syringe filters combine the benefits of a multi-layered prefiltration device with an automation-certified filter housing.
  • Increased surface area for maximizing throughput
  • Heat sealed membrane for reduced breakthrough
  • Accurate analysis. HPLC certified for low levels of UV-absorbing extractables
  • Available in 0.2 µm for UHPLC applications and with GxF (Glass Fiber) pre-filter for difficult to filter samples
  • Automation Certified to ensure smooth operation and worry-free performance 24 hours a day in automated
More Product Information