Ultra-Low Temperature Freezers, TSX Universal −86 °C ULT
Supplier: Thermo Fisher Scientific
TSX40086FARAKLB TSX40086FARAKLN TSX40086LA TSX50086FA TSX50086FALB TSX50086FARAK TSX50086FARAKCO TSX50086FARAKHID TSX50086FARAKLB TSX50086FARAKLN TSX40086FA TSX40086FALB TSX40086FARAK TSX40086FARAKCO TSX40086FARAKHID TSX50086LA TSX60086FA TSX60086FALB TSX60086FARAK TSX60086FARAKCO TSX60086FARAKHID TSX60086FARAKLB TSX60086FARAKLN TSX60086LA TSX70086FA TSX70086FALB TSX70086FARAK TSX70086FARAKCO TSX70086FARAKHID TSX70086FARAKLB TSX70086FARAKLN TSX70086LA TSX50086FARAKCO TSX50086FARAKLB TSX50086FARAKLN TSX50086FARAKHID TSX60086FA TSX60086FALB TSX60086LA TSX60086FARAK TSX60086FARCO TSX60086FARLB TSX60086FARLN TSX60086FARHID TSX60086FARAKCO TSX60086FARAKLB TSX60086FARAKLN TSX60086FARAKHID TSX70086FA TSX40086FARAKLN TSX40086FARAKHID TSX50086FA TSX50086FALB TSX50086LA TSX50086FARAK TSX50086FARCO TSX50086FARLB TSX50086FARLN TSX50086FARHID TSX70086FARCO TSX70086FARLB TSX70086FARLN TSX70086FARHID TSX70086FARAKCO TSX70086FARAKLB TSX70086FARAKLN TSX70086FARAKHID TSX40086FA TSX40086FALB TSX40086LA TSX40086FARAK TSX40086FARCO TSX40086FARLB TSX40086FARLN TSX40086FARHID TSX40086FARAKCO TSX40086FARAKLB TSX70086FALB TSX70086LA TSX70086FARAK
77400-782EA 39795.46 CAD
77400-782 77400-784 77400-786 77400-788 77400-790 77400-792 77400-794 77400-796 77400-798 77400-800 77400-772 77400-774 77400-776 77400-778 77400-780 77400-802 77400-804 77400-806 77400-808 77400-810 77400-812 77400-814 77400-816 77400-818 77400-820 77400-822 77400-824 77400-826 77400-828 77400-830 77400-832 77400-834 CA77400-794 CA77400-798 CA77400-800 CA77400-796 CA77400-804 CA77400-806 CA77400-818 CA77400-808 CA77603-424 CA77603-428 CA77603-430 CA77603-426 CA77400-810 CA77400-814 CA77400-816 CA77400-812 CA77400-820 CA77400-784 CA77400-780 CA77400-788 CA77400-790 CA77400-802 CA77400-792 CA77603-416 CA77603-420 CA77603-422 CA77603-418 CA77603-432 CA77603-436 CA77603-438 CA77603-434 CA77400-826 CA77400-830 CA77400-832 CA77400-828 CA77400-772 CA77400-774 CA77400-786 CA77400-776 CA77603-408 CA77603-412 CA77603-414 CA77603-410 CA77400-778 CA77400-782 CA77400-822 CA77400-834 CA77400-824
Ultra-Low Temperature Freezers, TSX Universal −86 °C ULT
Freezers Ultra Low Temperature Freezers
TSX Universal series ULT freezers are the pinnacle of reliability, sustainability, and innovation incorporating insights from over 80 years of experience to enhance every aspect of the system. Updates to the variable speed compressors has made the TSX Universal adaptable across all workflows. Available in four capacities, they are ENERGY STAR® certified with a 33% energy efficiency improvement, ACT labeled and produced in a Zero-Waste to Landfill certified facility.
  • Universal V-Drive technology delivers tighter peak variation, faster recovery times,
More Product Information