GC/MS for Advanced Battery Science Systems
Supplier: AGILENT TECHNOLOGIES, INC (VAD) CA
G7010CA G7012CA G7076CA G7077CA G7079CA G7250AA
CA77590-300EA 367259.05 CAD
CA77590-300 CA77590-302 CA77590-304 CA77590-306 CA77590-308 CA77590-310
GC/MS for Advanced Battery Science Systems
Spectrophotometers and Photometers Mass Spectrometers
The 5977C GC/MSD is a routine and reliable workhorse for environmental impurities and food testing, chemical and petrochemical analysis, as well as the analysis of forensic and pharmaceutical compounds.
The 7000E triple quadrupole GC/MS provides unequivocal intelligence and robustness to deliver the answers you seek. The routine workhorse for labs across a wide range of industry applications like pesticides in food or SVOCs in environmental samples now has SWARM autotune and improved diagnostic tools.
The 7010C GC triple quadrupole mass spectrometry system is the most sensitiveMore Product Information