Spectrometer ICP-MS for Advanced Battery Science Systems
Supplier: AGILENT TECHNOLOGIES, INC (VAD) CA
G3665AA G8403AA G8422AA
CA77590-294EA 552002.1 CAD
CA77590-294 CA77590-318 CA77590-320
Spectrometer ICP-MS for Advanced Battery Science Systems
Spectrophotometers and Photometers Mass Spectrometers
Free your ICP-MS analysis from common time traps with the Agilent 7850 ICP mass spectrometer. It’s the smart way to reduce wasted time so busy staff can focus on tasks that deliver value.​ The 7850 ICP-MS is an ICP mass spectrometer that can handle samples with up to 25% solids, reducing the dilution time trap. The instrument features a helium mode collision cell and half mass correction that remove both polyatomic and doubly charged ion interferences, making method development simpler and addressing a common cause of time-wasting sample remeasurement.
The 7900 ICP-MS is a flexible singleMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable