Cellulose Extraction Thimbles, Grade C300
Supplier: Sartorius
FT-1201-028100 FT-1201-030080 FT-1201-030100 FT-1201-033060 FT-1201-033080 FT-1201-033090 FT-1201-033094 FT-1201-033100 FT-1201-033118 FT-1201-033130 FT-1201-019090 FT-1201-022080 FT-1201-022100 FT-1201-025060 FT-1201-025070 FT-1201-025080 FT-1201-025100 FT-1201-026060 FT-1201-028060 FT-1201-028080 FT-1201-033205 FT-1201-035150 FT-1201-040100 FT-1201-040123 FT-1201-040150 FT-1201-043123
CA77581-162PK 249.08 CAD
CA77581-162 CA77581-164 CA77581-166 CA77581-168 CA77581-170 CA77581-172 CA77581-174 CA77581-176 CA77581-178 CA77581-180 CA77581-142 CA77581-144 CA77581-146 CA77581-148 CA77581-150 CA77581-152 CA77581-154 CA77581-156 CA77581-158 CA77581-160 CA77581-182 CA77581-184 CA77581-186 CA77581-188 CA77581-190 CA77581-192
Cellulose Extraction Thimbles, Grade C300
Extractors Extraction Thimbles
These cellulose thimble are primarily designed to fit the Soxhlet apparatus and potential applications include environmental monitoring (e.g. separation of dust, aerosols, gas or air streams) and food control (e.g. extraction of fats, emulsifiers or additives).
  • Composed of high-purity cellulose for uncontaminated results
  • Engineered for superior mechanical stability to withstand rigorous use
  • Designed for a precise fit across a diverse range of extractors
  • Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes