Real-Time PCR Thermal Cyclers, qTOWER iris, 115 V

Supplier: Analytik Jena CA
844-00853-4 844-00854-4 844-00855-4 844-00856-4 844-00858-4 844-00859-4
CA77497-288EA 47545 CAD
CA77497-288 CA77497-292 CA77497-296 CA77497-300 CA77497-304 CA77497-308
Real-Time PCR Thermal Cyclers, qTOWER iris, 115 V
Thermal Cyclers
The real-time PCR thermal cycler qTOWER iris empowers your exploration. It equips you to meet your current and future needs in qPCR. The instrument can hit six targets at once and is designed for long-term use with quiet operation and provides uniform precision across every well.

  • 115 V model
  • The complete spectrum: Clear signals from UV-A to Near Infrared (NIR)
  • Multiplexing: up to six targets simultaneously
  • Temperature accuracy ±0.15 °C
  • Dynamic range: 10 orders of magnitude
  • Your choice: Modules, consumables, reagents, and assays
  • Ergonomic and whisper-quiet
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR