Real-Time PCR Thermal Cyclers, qTOWER iris, 230 V

Supplier: Analytik Jena CA
844-00853-2 844-00854-2 844-00855-2 844-00856-2 844-00858-2 844-00859-2
CA77497-286EA 45716.4 CAD
CA77497-286 CA77497-290 CA77497-294 CA77497-298 CA77497-302 CA77497-306
Real-Time PCR Thermal Cyclers, qTOWER iris, 230 V
Thermal Cyclers
The real-time PCR thermal cycler qTOWER iris empowers your exploration. It equips you to meet your current and future needs in qPCR. The instrument can hit six targets at once and is designed for long-term use with quiet operation and provides uniform precision across every well.

  • 230 V model
  • The complete spectrum: Clear signals from UV-A to Near Infrared (NIR)
  • Multiplexing: up to six targets simultaneously
  • Temperature accuracy ±0.15 °C
  • Dynamic range: 10 orders of magnitude
  • Your choice: Modules, consumables, reagents, and assays
  • Ergonomic and whisper-quiet
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR