AssureTouch™ Protect Nitrile Medical Examination Gloves, Powder-Free
Supplier: Medicom
9990-B 9990-C 9990-D 9990-E 9990-A
CA77449-150BX 10.2 CAD
CA77449-150 CA77449-152 CA77449-154 CA77449-156 CA77449-148
AssureTouch™ Protect Nitrile Medical Examination Gloves, Powder-Free
Gloves
Non sterile, single use, medical examination gloves are intended to prevent transmission of a wide variety of diseases to both patients and health care personnel. Specifically, gloves are intended to act as a barrier between the health care provider and patients to prevent contamination between patient and caregiver.
  • Textured fingertips
  • Chemo-tested
  • Optimal tactile sensitivity
  • Comfortable fit: Nitrile gloves conform to hands, providing a snug, comfortable fit and reducing hand fatigue
  • Optimal stretch: Nitrile offers optimal flexibility, making nitrile
More Product Information