1M Sodium Phosphate Buffer
Supplier: TEKNOVA CA
P3072
CA77442-714EA 79.52 CAD
CA77442-714
1M Sodium Phosphate Buffer
Buffers Biological Buffers
312 mM sodium phosphate monobasic, 688 mM sodium phosphate dibasic.