Rapid Response™ Xylazine Test Strip Kit (Liquid/Powder), BTNX
Supplier: BTNX INC.
XYL-18S2-5K XYL-18S2-5K
77394-660EA 9.7 CAD
77394-660 CA77394-660
Rapid Response™ Xylazine Test Strip Kit (Liquid/Powder), BTNX
Drug Tests
The Rapid Response™ Xylazine Test Strip Kit (Liquid / Powder) detects the presence xylazine in liquid or powder samples at a cut-off concentration of 1000 ng/mL or higher.
  • Sample: Liquid/Powder
  • Detects xylazine at concentrations of 1000 ng/mL and above
  • Results as soon as 60 seconds
  • Full kit with all materials required to test drug samples
  • Results interpretation guide printed on each pouch
  • Easy to interpret results with built-in procedural controls
  • Individually packaged strips
Warning: This test does not evaluate drug safety or purity.
The RapidMore Product Information