Analytical Balances, Advanced MX
Supplier: Mettler Toledo
30665094 30665097 30665140 30665143 30665146 30665094 30665097 30665140 30665143 30665146 30665091
CA77323-040EA 15276.4 CAD
CA77323-040 CA77323-044 CA77323-048 CA77323-052 CA77323-056 77323-040 77323-044 77323-048 77323-052 77323-056 CA77393-646
Analytical Balances, Advanced MX
Balances and Scales
METTLER TOLEDO Advanced MX analytical balances offer a brand new feature- the backlit draft shield. Brightness extends over the draft shield's entire back panel providing exceptional visibility of the work area for highly precise dosing.
  • ErgoDoor
  • User management
  • Leveling assistant
  • StatusLight
  • Three interfaces: USB-A, USB-C, Ethernet
  • Bluetooth option
  • EasyDirect software
  • Full metal housing with overload protection
The stable SmartPan and high resolution weigh cell and FACT automatic internal adjustment ensure consistently accurate results. EfficientMore Product Information