Ocean Pacific® Elements Biodegradable Nitrile Medical Examination Gloves, Powder-Free
Supplier: Medicom
OPL-BIO OPM-BIO OPS-BIO OPXL-BIO OPXS-BIO
CA77322-270BX 23.97 CAD
CA77322-270 CA77322-268 CA77322-266 CA77322-272 CA77322-264
Ocean Pacific® Elements Biodegradable Nitrile Medical Examination Gloves, Powder-Free
Gloves
Non sterile, single use, medical examination gloves are intended to prevent transmission of a wide variety of diseases to both patients and health care personnel. Specifically, gloves are intended to act as a barrier between the health care provider and patients to prevent contamination between patient and caregiver.
  • Textured fingertips
  • Chemo and Fentanyl safe
  • 81% biodegradable in less than 1.5 years, 30% Biodegradation in 202 days
  • Specially formulated to speed up the natural biodegradation in landfills, without compromise to product quality and integrity
More Product Information