Upright Microscope with FITC Filter Fluorescence Bundle, Panthera C2
Supplier: Motic
1100104601331
CA77147-440EA 5282.99 CAD
CA77147-440
Upright Microscope with FITC Filter Fluorescence Bundle, Panthera C2
Microscopes
This new fluorescence bundle includes a Panthera C2 trinocular 50:50 microscope and a FITC filter (fluorescence accessory). If it is about stained sections with cover glass protection, about native smears, fine needle biopsies, blood smears which wanted to be treated with or without cover glass: Panthera C2 is your solution.
  • Bright field is the standard, phase contrast, pol contrast, objectives with/without cover glass correction, optional LED fluorescence modules with variable filter combinations
  • The new Plan UC Achromatic objectives, from 2× to 100×
More Product Information