Inverted Microscope with FITC and TRITC Filter Fluorescence Bundle, AE31E
Supplier: Motic
1100100200842
CA77147-436EA 16501.07 CAD
CA77147-436
Inverted Microscope with FITC and TRITC Filter Fluorescence Bundle, AE31E
Microscopes
This new fluorescence bundle includes AE31E LED trinocular inverted microscope, CCIS Plan Achromat phase LWD objective, universal mechanical stage, 3 size of petri dish holder, complete EPI-fluorescence attachment, EGFP/FITC/Cy2/AlexaFluor 488 and TRITC/Cy3/TagRFP/AlexaFluor 546. This is the perfect platform for all kind of routine microbiology applications in clinical and pharmaceutical laboratories.
  • Display maximum functionality and ergonomics in all its mechanical aspects
  • Elegant and robust, the microscope stand follows all needs for an intense daily
More Product Information