Hei-PLATE Mix 'N' Heat Expert Magnetic Hotplate Stirrers
Supplier: Heidolph NA, LLC
036110230CA 036110231CA 036110234CA 036110235CA 036110236CA 036110237CA
CA76777-166EA 2353.86 CAD
CA76777-166 CA76777-168 CA76777-174 CA76777-176 CA76777-178 CA76777-180
Hei-PLATE Mix 'N' Heat Expert Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
For smooth to intense mixing and heating of low-viscosity fluids the Hei-PLATE Mix 'N' Heat Expert magnetic hotplate stirrers, are an excellent choice for use with decomposing organic and inorganic substances.
  • One display for each main function (stirring and heating) – provides clear view of all values and settings at any time
  • Set parameters via 2 push-and-rotate controls with LED-rings for easy identification of active functions
  • Separate timers for heating and stirring functions mean flexibility and improved control of experiments
  • Choice of 3 heating
More Product Information