High Sensitivity DNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
5067-5592 5067-5593 5067-5594 5067-5603 5067-5584 5067-5585 5067-5587
CA76645-084EA 632.94 CAD
CA76645-084 CA76645-492 CA76645-062 CA76645-494 CA76645-162 CA76645-270 CA76645-118
High Sensitivity DNA ScreenTape Analysis
Electrophoresis Systems Capillary Electrophoresis Systems
DNA fragment or library analysis from samples of limited abundance, for samples up to 5000 bp.
  • The broad size range allows analysis of DNA fragments or libraries from 35 to 5000 bp
  • Results are obtained quickly, in approximately 1 minute per sample
  • Detection of DNA samples with limited abundance is possible, for samples down to 5 pg/µl
  • Ready-to use ScreenTape devices, pre-packaged reagent and automated data analysis offer an easy to use system with reproducible DNA sizing and quantification results
  • Reliable sample quality control is provided for next-
More Product Information