Small Fragment and NGS Kits
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
DNF-230-0240 DNF-240-0240 DNF-240-0500 DNF-473-0500 DNF-473-1000 DNF-474-0500 DNF-474-1000 DNF-476-0500 DNF-477-0500
CA76645-132EA 1350.32 CAD
CA76645-132 CA76645-236 CA76645-408 CA76645-324 CA76645-098 CA76645-304 CA76645-046 CA76645-156 CA76645-250
Small Fragment and NGS Kits
Electrophoresis Systems Capillary Electrophoresis Systems
Quality control for DNA smears and fragments including cfDNA analysis and NGS library QC.
  • Smear concentration range - The HS NGS and HS small fragment kits have a smear concentration range of 50 to 5000 pg/µl, The NGS and small fragment kits covers a higher range of 5 to 100 ng/µl
  • Fragment concentration range - The HS NGS and HS small fragment kits have a fragment concentration range of 5 to 500 pg/µl, The NGS and small fragment kits covers a higher input range of 0.1 to 10 ng/µl
  • Reliable sizing - kits for small and standard sized DNA fragments and smears
More Product Information