SCIEX Triple Quad 7500 Systems
Supplier: AB SCIEX LLC CA
5069160 5077299 5069160 5077299
76644-992EA 855926.76 CAD
76644-992 76644-996 CA76644-992 CA76644-996
SCIEX Triple Quad 7500 Systems
Spectrophotometers and Photometers Mass Spectrometers
The SCIEX 7500 system delivers a higher level of sensitivity than previous SCIEX systems. The system is integrated with innovative technology, which enables a low-level of quantification of analytes in crude and complex matrices.
  • QTRAP activated and QTRAP ready options
  • The D Jet ion guide concentrates samples and removes gas molecules and neutral ions
  • Enhanced product ion scans enable improved confidence, while MRM3 workflows push quantification levels through matrix interferences
  • Source: OptiFlow Pro ion source housing with OptiFlow Pro analytical probe
More Product Information