Rapid Response™ Xylazine Test Strip (Liquid/Powder), BTNX
Supplier: BTNX INC.
XYL-18S2-100 XYL-18S2-10 XYL-18S2-100 XYL-18S2-10
76642-512EA 489.42 CAD
76642-512 77394-658 CA76642-512 CA77394-658
Rapid Response™ Xylazine Test Strip (Liquid/Powder), BTNX
Drug Tests
The Rapid Response™ Xylazine Test Strip (Liquid / Powder) detects the presence xylazine in liquid or powder samples at a cut-off concentration of 1000 ng/mL or higher.
  • Sample: Liquid/Powder
  • Quantity: 100 Tests/Kit
  • Detects xylazine at concentrations of 1000 ng/mL and above
  • Results as soon as 60 seconds
  • Results interpretation guide printed on each pouch
  • Easy to interpret results with built-in procedural controls
  • Individually packaged strips for easy distribution
Warning: This test does not evaluate drug safety or purity.
The Rapid Response™ Xylazine TestMore Product Information