Mica Widefield and Widefocal Microscopes, Leica Microsystems
Supplier: Leica Microsystems
11889183 11506517 11506535 11513859 11525700 11525712 11525717 11600220 11889180 9I_CM_APPLIC1 9I_CM_MICA_B 9I_CM_MICA_D
CA76575-548EA 128328.85 CAD
CA76575-548 CA76635-926 CA76635-928 CA76635-930 CA76635-932 CA76635-934 CA76635-936 CA76635-938 CA76635-940 CA76635-942 CA76635-944 CA76635-946 CA76635-948 CA76635-950
Mica Widefield and Widefocal Microscopes, Leica Microsystems
Microscopes
More than a highly automated microscope, Mica unites widefield and confocal imaging in a sample protecting environment. With the simple push of a button, you have everything you need all in one place to supercharge fluorescence imaging workflows and get meaningful scientific results faster.
  • Now you can focus on your science, not figuring out your microscope
  • Access for all
  • No constraints
  • Radically simplified workflows
Mica provides a clear sample overview and allows to easily change observation conditions with just a few clicks. The Microhub enablesMore Product Information