Whatman™ Pyramid Folded Filter Papers, Whatman products (Cytiva)
Supplier: Cytiva
1001-10116 1001-10218 1002-10219
CA76635-522PK 2917.58 CAD
CA76635-522 CA76635-535 CA76635-527
Whatman™ Pyramid Folded Filter Papers, Whatman products (Cytiva)
Filters Filter Papers Quantitative Filter Paper
Whatman cellulose filter paper grades are also available in pyramid (square) folded formats.
  • Easily fits into conical funnels
  • Saves valuable time when undertaking repetitive or multiple analyses
  • Provides the same high quality and consistency that you expect from all Whatman filter papers
Order Now
SPECIFICATIONS
Environmentally Preferable
Ash content - 0.06% 0.06%