HiTrap™ Capto Core™ 700 Multimodal Chromatography Columns
Supplier: Cytiva
29400461
CA76633-048EA 117.76 CAD
CA76633-048
HiTrap™ Capto Core™ 700 Multimodal Chromatography Columns
Chromatography Columns
HiTrap™ Capto Core 700 is prepacked with Capto Core 700, a multimodal BioProcess medium for intermediate purification and polishing of viruses and other large biomolecules.
  • High capacity and productivity in flowthrough mode
  • Wide operational window of pH and conductivity
  • Prepacked HiTrap™ columns for easy screening and convenient process development, as well as small scale purification
Capto Core 700 is a novel, layered bead chromatography resin consisting of a porous outer layer and ligand-activated core. The octylamine ligands found in the core ofMore Product Information