Racks for Freezers
Supplier: Eppendorf
6001000310 6001000311 6001000410 6001000411 6001000510 6001000910 6001000911 6001011110 6001011210 6001011310 6001011410 6001011510 6001011910 6001012210 6001012310 6001012410 6001012510 6001012910 6001021110 6001021210 6001021310 6001021410 6001021910 6001022210 6001022310 6001022410 6001022910 6001040010 6001040011 6001040110 6001040111 6001040210 6001040211 6001050010 6001050011 6001050110 6001050111 6001050210 6001050211 6001061210 6001061310 6001061410 6001061510 6001061910 6001062210 6001062310 6001062410 6001062510 6001062910 6001071110 6001071210 6001071310 6001071410 6001071510 6001071910 6001072210 6001072310 6001072410 6001072510 6001072910 6001081110 6001081210 6001081310 6001081410 6001081510 6001081910 6001082210 6001082310 6001082410 6001082510 6001082910
CA76545-676EA 251.31 CAD
CA76545-676 CA76545-678 CA76545-680 CA76545-682 CA76545-684 CA76545-686 CA76545-688 CA76545-690 CA76545-692 CA76545-694 CA76545-696 CA76545-698 CA76545-700 CA76545-702 CA76545-704 CA76545-706 CA76545-708 CA76545-710 CA76545-712 CA76545-714 CA76545-716 CA76545-718 CA76545-720 CA76545-722 CA76545-724 CA76545-726 CA76545-728 CA76545-730 CA76545-732 CA76545-734 CA76545-736 CA76545-738 CA76545-740 CA76545-742 CA76545-744 CA76545-746 CA76545-748 CA76545-750 CA76545-752 CA76545-754 CA76545-756 CA76545-758 CA76545-760 CA76545-762 CA76545-764 CA76545-766 CA76545-768 CA76545-770 CA76545-772 CA76545-774 CA76545-776 CA76545-778 CA76545-780 CA76545-782 CA76545-784 CA76545-786 CA76545-788 CA76545-790 CA76545-792 CA76545-794 CA76545-796 CA76545-798 CA76545-800 CA76545-802 CA76545-804 CA76545-806 CA76545-808 CA76545-810 CA76545-812 CA76545-814 CA76545-816
Racks for Freezers
Racks Cryogenic Box Racks
These racks are heavy-duty and designed for higher capacity and durability. Specially designed for optimised cooling conditions and improved sample safety.
  • Fit freezer boxes up to a 136×136 mm for increased storage flexibility
  • Drawer-style freezer racks, each drawer can be individually removed, leaving the rest in the freezer
  • In side-access racks each box fits into a distinct, individual compartment for easier inventory tracking
There are two systems used in laboratories - the standard 4-boxes deep racks and 5-boxes deep racks. The easiest way is toMore Product Information