BD PhaSeal™ Optima System, BD Medical
Supplier: BD
515052 515060 515064 515070 515078
CA76545-554CS 2429.48 CAD
CA76545-554 CA76545-576 CA76545-558 CA76545-572 CA76545-556
BD PhaSeal™ Optima System, BD Medical
Infusions
Various components of a closed-system drug transfer device (CSTD) solution for hazardous drug protection. The BD PhaSeal™ Optima system meets the requirements of the NIOSH IPA draft CSTD-test protocol, a robust test protocol applicable to barrier-type closed system drug transfer devices.
  • Every component of the BD PhaSeal Optima System is designed with no inlets or air exchange for airtight hazardous drug transfers
  • PhaSeal™ Optima System is leakproof for both short- and long-term connections
  • Membranes self-seal upon disconnect
The BD PhaSeal™More Product Information