SafeMask® FreeFlow™ Procedure Earloop Face Masks, Medicom
Supplier: Medicom
200311 200315 200316 200415 200416 200511 200514 200515 200516
CA76544-748CS 198.79 CAD
CA76544-748 CA76544-750 CA76544-752 CA76544-754 CA76544-756 CA76544-758 CA76544-760 CA76544-762 CA76544-764
SafeMask® FreeFlow™ Procedure Earloop Face Masks, Medicom
Masks Surgical Masks
Breathe easy, see clearly, and stay comfortable with the SafeMask® FreeFlow™ Procedure Earloop Face Masks.
  • Non-woven spunbond outer layer provides maximum protection against fluid penetration
  • Masks feature aluminum nose- piece, flat knitted earloop, and anti-fog strip
  • Omega pleats create a unique air pocket for breathability
  • High-quality media ensures optimal filtration and breathability
  • Adjustable, enclosed nosepiece forms a strong seal to provide customized protection and comfort
  • Latex-free
Innovative contoured pleats create a unique pocket thatMore Product Information