PUREZERO* HG3 White SGX* Nitrile Gloves, Halyard
Supplier: O&M HALYARD CANADA, INC CA
CLN3231LG CLN3231MD CLN3231SM CLN3231XL CLN3231XS
CA76540-260CS 519.34 CAD
CA76540-260 CA76540-262 CA76540-264 CA76540-266 CA76540-268
PUREZERO* HG3 White SGX* Nitrile Gloves, Halyard
Gloves
PUREZERO* Cleanroom Gloves are ideal for applications that involve handling delicate equipment in microelectronics, semiconductors, optics, pharmaceutical, and medical device manufacturing applications. They are specifically designed to meet the stringent requirements of cleanroom environments.
  • Recommended for use in ISO Class 3 or higher and Grade A/B/C/D cleanrooms
  • Maximum particles: >0.5 µm/cm² less than 950
  • AQL 0.65 for pinholes
  • Accelerator-, silicone-, latex-, and powder-free
  • 30.5 cm (12") length with beaded cuff
  • Fingertip thickness: 0.16 mm/6 mil
More Product Information